Anti-Perforin Mouse Monoclonal Antibody [clone: dG9] (Brilliant Violet® 711)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG308129
- BLEG308130
- Antigen symbol:
- Perforin
- Perforin
- Antigen synonyms:
- P1|PRF1|PFP|HPLH2
- P1|PRF1|PFP|HPLH2
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Brilliant Violet® 711
- Brilliant Violet® 711
- Clone:
- dG9
- dG9
- Reactivity:
- Human, Bovine
- Human, Bovine
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Purified granules from the human lymphoma cell line
- Purified granules from the human lymphoma cell line
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711™ under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711™ under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- ICFC
- ICFC
Specifications
Frequently Bought Together