Anti-CD357 (GITR) Mouse Monoclonal Antibody [clone: 108-17] (Brilliant Violet® 711)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG371212
- BLEG371211
- Antigen symbol:
- CD357 (GITR)
- CD357 (GITR)
- Antigen synonyms:
- Glucocorticoid-inducible TNFR|TNFRSF18|AITR|Activation-inducible TNFR
- Glucocorticoid-inducible TNFR|TNFRSF18|AITR|Activation-inducible TNFR
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Brilliant Violet® 711
- Brilliant Violet® 711
- Clone:
- 108-17
- 108-17
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Recombinant human GITR-Fc chimera
- Recombinant human GITR-Fc chimera
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711™ under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711™ under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Specifications
Frequently Bought Together