Anti-CD81 (TAPA-1) Mouse Monoclonal Antibody [clone: 5A6] (Pacific Blue™)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG349515
- BLEG349516
- Antigen symbol:
- CD81 (TAPA-1)
- CD81 (TAPA-1)
- Antigen synonyms:
- S5.7|TSPAN28|CVID6
- S5.7|TSPAN28|CVID6
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Pacific Blue™
- Pacific Blue™
- Clone:
- 5A6
- 5A6
- Reactivity:
- Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
- Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Human OCI-LY8 cell line
- Human OCI-LY8 cell line
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Pacific Blue™ under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Pacific Blue™ under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Specifications
Frequently Bought Together