Anti-T-bet Mouse Monoclonal Antibody [clone: 4B10] (Brilliant Violet® 421)
Catalog # BLEG644832
Anbieter: Biolegend
Technische Daten
- Antigen symbol:T-bet
- Clonality:Monoclonal
- Clone:4B10
- Conjugation:Brilliant Violet® 421
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Antigen synonyms:T-box expressed in T cells|T box 21|TBLYM
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:2…8 °C
- Concentration:0.2 mg/ml
- Tested applications:ICFC
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions.
- Size:50 µg
- Pk:50 µG