Anti-CD172a (SIRPalpha) Rat Monoclonal Antibody [clone: P84] (Alexa Fluor® 700)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG144021
- BLEG144022
- Antigen symbol:
- CD172a (SIRPalpha)
- CD172a (SIRPalpha)
- Antigen synonyms:
- CD172 antigen-like family member A|PTPNS1|SHPS-1|P84|BIT
- CD172 antigen-like family member A|PTPNS1|SHPS-1|P84|BIT
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 700
- Alexa Fluor® 700
- Clone:
- P84
- P84
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Mouse brain membrane protein
- Mouse brain membrane protein
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 700 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 700 under optimal conditions.
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Specifications
Frequently Bought Together