Anti-TIGIT (VSTM3) Mouse Monoclonal Antibody [clone: A15153G] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG372717
- BLEG372718
- Antigen symbol:
- TIGIT (VSTM3)
- TIGIT (VSTM3)
- Antigen synonyms:
- WUCAM|VSTM3|T-cell immunoreceptor with Ig and ITIM domains|VSIG9
- WUCAM|VSTM3|T-cell immunoreceptor with Ig and ITIM domains|VSIG9
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Clone:
- A15153G
- A15153G
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Recombinant Human TIGIT.
- Recombinant Human TIGIT.
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PerCP/Cyanine5.5 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PerCP/Cyanine5.5 under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Specifications
Frequently Bought Together