Anti-CD152 (CTLA-4) Mouse Monoclonal Antibody [clone: BNI3] (APC (Allophycocyanin))
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG369611
- BLEG369612
- Antigen symbol:
- CD152 (CTLA-4)
- CD152 (CTLA-4)
- Antigen synonyms:
- Cytotoxic T-Lymphocyte Antigen 4|CTLA4
- Cytotoxic T-Lymphocyte Antigen 4|CTLA4
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- BNI3
- BNI3
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Extracellular domain of human CTLA-4 and constant regions of the human IgG heavy chain (CTLA-4/IgG)
- Extracellular domain of human CTLA-4 and constant regions of the human IgG heavy chain (CTLA-4/IgG)
- Purification:
- The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- ICFC, FC
- ICFC, FC
Specifications
Frequently Bought Together