Anti-CD34 Mouse Monoclonal Antibody [clone: 561] (Brilliant Violet® 650)
- Catalog No:
- BLEG343624
- BLEG343623
- Antigen symbol:
- CD34
- CD34
- Antigen synonyms:
- Mucosialin|Gp105-120|My10
- Mucosialin|Gp105-120|My10
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Brilliant Violet® 650
- Brilliant Violet® 650
- Clone:
- 561
- 561
- Reactivity:
- Cynomolgus monkey, Rhesus monkey, Human
- Cynomolgus monkey, Rhesus monkey, Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow cytometry:
- Yes
- Yes
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 650™ under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 650™ under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Frequently Bought Together