Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (Brilliant Violet® 650)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG302946
- BLEG302945
- Antigen symbol:
- CD28
- CD28
- Antigen synonyms:
- Tp44|T44
- Tp44|T44
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Brilliant Violet® 650
- Brilliant Violet® 650
- Clone:
- CD28.2
- CD28.2
- Reactivity:
- Baboon, Capuchin monkey, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human, Squirrel monkey
- Baboon, Capuchin monkey, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human, Squirrel monkey
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 650™ under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 650™ under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Specifications
Frequently Bought Together