Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD303 (BDCA-2) Mouse Monoclonal Antibody [clone: 201A] (Brilliant Violet® 605)
Anti-CD303 (BDCA-2) Mouse Monoclonal Antibody [clone: 201A] (Brilliant Violet® 605)
Supplier:  Biolegend
Certificates
Anti-CD303 (BDCA-2) Mouse Monoclonal Antibody [clone: 201A] (Brilliant Violet® 605)
Supplier:  Biolegend

Specifications

  • Catalog No:
  • BLEG354223
  • BLEG354224
  • Antigen symbol:
  • CD303 (BDCA-2)
  • CD303 (BDCA-2)
  • Antigen synonyms:
  • CLECSF11|CLEC4C|HECL|CLECSF7|DLEC
  • CLECSF11|CLEC4C|HECL|CLECSF7|DLEC
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Brilliant Violet® 605
  • Brilliant Violet® 605
  • Clone:
  • 201A
  • 201A
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 605™ under optimal conditions.
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 605™ under optimal conditions.
  • Storage buffer:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Storage temperature:
  • 2…8 °C
  • 2…8 °C
  • Tested applications:
  • FC
  • FC

Specifications

Frequently Bought Together