Anti-CD20 Rat Monoclonal Antibody [clone: SA275A11] (PE (Phycoerythrin))
Nome del fornitore: Biolegend
Certificati
Informazioni su questo articolo
Specifiche
- Catalog No:
- BLEG150409
- BLEG150410
- Antigen symbol:
- CD20
- CD20
- Antigen synonyms:
- Ly-44|Ms4a1
- Ly-44|Ms4a1
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- SA275A11
- SA275A11
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Mouse CD20 - transfected cells
- Mouse CD20 - transfected cells
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- Concentration:
- 0.2 mg/ml
- 0.2 mg/ml
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Specifiche
Frequently Bought Together