Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD27 Mouse Monoclonal Antibody [clone: M-T271] (APC (Allophycocyanin)/Cy7®)
Anti-CD27 Mouse Monoclonal Antibody [clone: M-T271] (APC (Allophycocyanin)/Cy7®)
Supplier:  Biolegend
Certificates
Anti-CD27 Mouse Monoclonal Antibody [clone: M-T271] (APC (Allophycocyanin)/Cy7®)
Supplier:  Biolegend

Specifications

  • Catalog No:
  • BLEG356423
  • BLEG356424
  • Antigen symbol:
  • CD27
  • CD27
  • Antigen synonyms:
  • S152|T14|TNFRSF7
  • S152|T14|TNFRSF7
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)/Cy7®
  • APC (Allophycocyanin)/Cy7®
  • Clone:
  • M-T271
  • M-T271
  • Reactivity:
  • Baboon, Cynomolgus monkey, Rhesus monkey, Human
  • Baboon, Cynomolgus monkey, Rhesus monkey, Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Human T cells from a T-ALL patient.
  • Human T cells from a T-ALL patient.
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with APC/Cyanine7 under optimal conditions.
  • The antibody was purified by affinity chromatography and conjugated with APC/Cyanine7 under optimal conditions.
  • Storage buffer:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Storage temperature:
  • 2…8 °C
  • 2…8 °C
  • Tested applications:
  • FC
  • FC

Specifications

Frequently Bought Together