Order Entry
Switzerland
ContactUsLinkComponent
Anti-Granzyme K Mouse Monoclonal Antibody [clone: GM26E7] (Alexa Fluor® 647)
Anti-Granzyme K Mouse Monoclonal Antibody [clone: GM26E7] (Alexa Fluor® 647)
Supplier:  Biolegend
Certificates
Anti-Granzyme K Mouse Monoclonal Antibody [clone: GM26E7] (Alexa Fluor® 647)
Supplier:  Biolegend

Specifications

  • Catalog No:
  • BLEG370503
  • BLEG370504
  • Antigen symbol:
  • Granzyme K
  • Granzyme K
  • Antigen synonyms:
  • Tryptase II|Granzyme-3|fragmentin-3|GZMK
  • Tryptase II|Granzyme-3|fragmentin-3|GZMK
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 647
  • Alexa Fluor® 647
  • Clone:
  • GM26E7
  • GM26E7
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 647 under optimal conditions.
  • The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 647 under optimal conditions.
  • Storage buffer:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Storage temperature:
  • 2…8 °C
  • 2…8 °C
  • Tested applications:
  • ICFC
  • ICFC

Specifications

Frequently Bought Together