Anti-CD45 Mouse Monoclonal Antibody [clone: 2D1] (APC (Allophycocyanin))
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG368512
- BLEG368511
- Antigen symbol:
- CD45
- CD45
- Antigen synonyms:
- T200|Leukocyte Common Antigen (LCA)
- T200|Leukocyte Common Antigen (LCA)
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- 2D1
- 2D1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Human PBMC
- Human PBMC
- Purification:
- The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Specifications
Frequently Bought Together