Anti-CD20 Mouse Monoclonal Antibody [clone: 2H7] (Brilliant Violet® 785)
- Catalog No:
- BLEG302356
- BLEG302355
- Antigen symbol:
- CD20
- CD20
- Antigen synonyms:
- Bp35|B1
- Bp35|B1
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Brilliant Violet® 785
- Brilliant Violet® 785
- Clone:
- 2H7
- 2H7
- Reactivity:
- Baboon, Capuchin monkey, Cynomolgus monkey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human, Squirrel monkey
- Baboon, Capuchin monkey, Cynomolgus monkey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human, Squirrel monkey
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Human tonsillar B cells
- Human tonsillar B cells
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 785™ under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 785™ under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Frequently Bought Together