Anti-IL-31 Mouse Monoclonal Antibody [clone: 1D10B31] (PE (Phycoerythrin))
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG659604
- BLEG659603
- Antigen-Symbol:
- IL-31
- IL-31
- Antigen-Synonyme:
- Interleukin-31
- Interleukin-31
- Klonalität:
- Monoklonal
- Monoklonal
- Konjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Klon:
- 1D10B31
- 1D10B31
- Reaktivität:
- Human
- Human
- Wirt:
- Mouse
- Mouse
- Isotyp:
- IgG2b kappa
- IgG2b kappa
- Durchflusszytometrie:
- Yes
- Yes
- Immunogen:
- Full length human IL-31 recombinant protein expressed in E. coli.
- Full length human IL-31 recombinant protein expressed in E. coli.
- Reinigung:
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- Lagerungspuffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Lagertemperatur:
- 2…8 °C
- 2…8 °C
- Validierte Anwendungen:
- ICFC
- ICFC
Specifications
Frequently Bought Together