Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD40 Mouse Monoclonal Antibody [clone: 5C3] (Brilliant Violet® 650)
Anti-CD40 Mouse Monoclonal Antibody [clone: 5C3] (Brilliant Violet® 650)
Supplier:  Biolegend
Certificates
Anti-CD40 Mouse Monoclonal Antibody [clone: 5C3] (Brilliant Violet® 650)
Supplier:  Biolegend

Specifications

  • Catalog No:
  • BLEG334338
  • BLEG334337
  • Antigen symbol:
  • CD40
  • CD40
  • Antigen synonyms:
  • BP50|TNFRSF5
  • BP50|TNFRSF5
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Brilliant Violet® 650
  • Brilliant Violet® 650
  • Clone:
  • 5C3
  • 5C3
  • Reactivity:
  • Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Squirrel monkey
  • Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Squirrel monkey
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 650™ under optimal conditions.
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 650™ under optimal conditions.
  • Storage buffer:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Storage temperature:
  • 2…8 °C
  • 2…8 °C
  • Tested applications:
  • FC
  • FC

Specifications

Frequently Bought Together