Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD66b Mouse Monoclonal Antibody [clone: G10F5] (Alexa Fluor® 700)
Anti-CD66b Mouse Monoclonal Antibody [clone: G10F5] (Alexa Fluor® 700)
Supplier:  Biolegend
Certificates
Anti-CD66b Mouse Monoclonal Antibody [clone: G10F5] (Alexa Fluor® 700)
Supplier:  Biolegend

Specifications

  • Catalog No:
  • BLEG305114
  • BLEG305113
  • Antigen symbol:
  • CD66b
  • CD66b
  • Antigen synonyms:
  • CEACAM8|NCA-95|CGM6|CD67
  • CEACAM8|NCA-95|CGM6|CD67
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 700
  • Alexa Fluor® 700
  • Clone:
  • G10F5
  • G10F5
  • Reactivity:
  • Chimpanzee, Human
  • Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgM kappa
  • IgM kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 700 under optimal conditions.
  • The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 700 under optimal conditions.
  • Storage buffer:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Storage temperature:
  • 2…8 °C
  • 2…8 °C
  • Tested applications:
  • FC
  • FC

Specifications

Frequently Bought Together