Order Entry
Switzerland
ContactUsLinkComponent
Anti-Ly-6C Rat Monoclonal Antibody [clone: HK1.4] (Brilliant Violet® 785)
Anti-Ly-6C Rat Monoclonal Antibody [clone: HK1.4] (Brilliant Violet® 785)
Catalog # BLEG128041
Supplier:  Biolegend
Anti-Ly-6C Rat Monoclonal Antibody [clone: HK1.4] (Brilliant Violet® 785)
Catalog # BLEG128041
Supplier:  Biolegend

Specifications

  • Antigen symbol:
    Ly-6C
  • Clonality:
    Monoclonal
  • Clone:
    HK1.4
  • Conjugation:
    Brilliant Violet® 785
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2c kappa
  • Reactivity:
    Mouse
  • Antigen synonyms:
    locus C|Lymphocyte antigen 6 complex
  • Storage buffer:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Storage temperature:
    2…8 °C
  • Concentration:
    0.2 mg/ml
  • Immunogen:
    L3 cloned CTL cells
  • Tested applications:
    FC
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 785™ under optimal conditions.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item