Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-CD62P (P-selectin) Mouse Monoclonal Antibody [clone: RMP-1] (APC (Allophycocyanin))
 :  Biolegend
 
Anti-CD62P (P-selectin) Mouse Monoclonal Antibody [clone: RMP-1] (APC (Allophycocyanin))
 :  Biolegend

 

  • Catalog No:
  • BLEG148303
  • BLEG148304
  • Antigen symbol:
  • CD62P (P-selectin)
  • CD62P (P-selectin)
  • Antigen synonyms:
  • GMP-140|Platelet activation-dependent granule membrane protein (PADGEM)
  • GMP-140|Platelet activation-dependent granule membrane protein (PADGEM)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • RMP-1
  • RMP-1
  • Reactivity:
  • Rat, Mouse
  • Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Thrombin activated rat platelets
  • Thrombin activated rat platelets
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions.
  • The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions.
  • Concentration:
  • 0.2 mg/ml
  • 0.2 mg/ml
  • Storage buffer:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Storage temperature:
  • 2…8 °C
  • 2…8 °C
  • Tested applications:
  • FC
  • FC

 

Frequently Bought Together