Anti-Nuclear Pore Complex Proteins Mouse Monoclonal Antibody [clone: MAb414]
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG902901
- BLEG902902
- Antigen symbol:
- Nuclear Pore Complex Proteins
- Nuclear Pore Complex Proteins
- Antigen synonyms:
- Nuclear pore complex protein Nup107|NUP98|Nucleoporin Nup107|NUP84|NPC proteins|NUP107|NUP 107|Nucleoporin 107kD|NUP153|NUP155|107 kD nucleoporin|NUP 84
- Nuclear pore complex protein Nup107|NUP98|Nucleoporin Nup107|NUP84|NPC proteins|NUP107|NUP 107|Nucleoporin 107kD|NUP153|NUP155|107 kD nucleoporin|NUP 84
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- MAb414
- MAb414
- Reactivity:
- Yeast, Xenopus, Vertebrates
- Yeast, Xenopus, Vertebrates
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Immunogen:
- The antibody was raised using a nuclear pore complex mixture.
- The antibody was raised using a nuclear pore complex mixture.
- Format:
- Purified
- Purified
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- Phosphate-buffered solution + 0.03% Thimerosal.
- Phosphate-buffered solution + 0.03% Thimerosal.
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- ICC, WB, IHC-F, IP, IEM
- ICC, WB, IHC-F, IP, IEM
Specifications
Frequently Bought Together