Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-GFP Epitope Tag Mouse Monoclonal Antibody [clone: B34]
 :  Biolegend
 
Anti-GFP Epitope Tag Mouse Monoclonal Antibody [clone: B34]
 :  Biolegend

 

  • Catalog No:
  • BLEG902601
  • BLEG902602
  • Antigen symbol:
  • GFP Epitope Tag
  • GFP Epitope Tag
  • Antigen synonyms:
  • Green Fluorescent Protein
  • Green Fluorescent Protein
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • B34
  • B34
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Immunogen:
  • Antibody B34, generated against purified wild type GFP, offers an alternitive for detecting GFP and GFP variants including enhanced/mutant GFP.
  • Antibody B34, generated against purified wild type GFP, offers an alternitive for detecting GFP and GFP variants including enhanced/mutant GFP.
  • Format:
  • Purified
  • Purified
  • Purification:
  • The antibody was purified by affinity chromatography.
  • The antibody was purified by affinity chromatography.
  • Concentration:
  • 1 mg/ml
  • 1 mg/ml
  • Storage buffer:
  • Phosphate-buffered solution + 0.03% Thimerosal.
  • Phosphate-buffered solution + 0.03% Thimerosal.
  • Storage temperature:
  • 2…8 °C
  • 2…8 °C
  • Tested applications:
  • WB, Purification
  • WB, Purification

 

Frequently Bought Together