Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-beta-Amyloid, 1-40 Mouse Monoclonal Antibody [clone: 11A50-B10] (Biotin)
  BLEG805404
 :  Biolegend
Anti-beta-Amyloid, 1-40 Mouse Monoclonal Antibody [clone: 11A50-B10] (Biotin)
  BLEG805404
 :  Biolegend

 

  • Antigen symbol:
    beta-Amyloid, 1-40
  • Clonality:
    Monoclonal
  • Clone:
    11A50-B10
  • Conjugation:
    Biotin
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    Azide-free
  • Antigen synonyms:
    APP|CTFgamma|CVAP|protease nexin-II|ABETA|cerebral vascular amyloid peptide|beta-amyloid peptide|APPI|AD1|PN-II|Amyloid Precursor Protein|preA4|ABPP|alzheimer disease amyloid protein|Amyloid beta A4 protein|AAA|PN2|peptidase nexin-II
  • Storage buffer:
    Phosphate-buffered solution; no preservatives or carrier proteins.
  • Storage temperature:
    2…8 °C
  • Concentration:
    0.5 mg/ml
  • Tested applications:
    ELISA, WB, IHC, IP
  • Purification:
    The antibody was purified by affinity chromatography.
  • Size:
    200 µl
  • Pk:
    200 µl