Anti-beta-Amyloid, 1-40 Mouse Monoclonal Antibody [clone: 11A50-B10]
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG805401
- BLEG805402
- BLEG805403
- Antigen symbol:
- beta-Amyloid, 1-40
- beta-Amyloid, 1-40
- beta-Amyloid, 1-40
- Antigen synonyms:
- APP|CTFgamma|CVAP|protease nexin-II|ABETA|cerebral vascular amyloid peptide|beta-amyloid peptide|APPI|AD1|PN-II|Amyloid Precursor Protein|preA4|ABPP|alzheimer disease amyloid protein|Amyloid beta A4 protein|AAA|PN2|peptidase nexin-II
- APP|CTFgamma|CVAP|protease nexin-II|ABETA|cerebral vascular amyloid peptide|beta-amyloid peptide|APPI|AD1|PN-II|Amyloid Precursor Protein|preA4|ABPP|alzheimer disease amyloid protein|Amyloid beta A4 protein|AAA|PN2|peptidase nexin-II
- APP|CTFgamma|CVAP|protease nexin-II|ABETA|cerebral vascular amyloid peptide|beta-amyloid peptide|APPI|AD1|PN-II|Amyloid Precursor Protein|preA4|ABPP|alzheimer disease amyloid protein|Amyloid beta A4 protein|AAA|PN2|peptidase nexin-II
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- 11A50-B10
- 11A50-B10
- 11A50-B10
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
- ELISA:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Format:
- Azide-free
- Azide-free
- Azide-free
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- 0.5 mg/ml
- Storage buffer:
- Phosphate-buffered solution (no preservatives or carrier proteins).
- Phosphate-buffered solution (no preservatives or carrier proteins).
- Phosphate-buffered solution (no preservatives or carrier proteins).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- 2…8 °C
- Tested applications:
- IHC-P, Dot Blot, ELISA
- IHC-P, Dot Blot, ELISA
- IHC-P, Dot Blot, ELISA
Specifications
Frequently Bought Together