Anti-beta-Amyloid, 17-24 Mouse Monoclonal Antibody [clone: 4G8]
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG800701
- BLEG800703
- Antigen symbol:
- beta-Amyloid, 17-24
- beta-Amyloid, 17-24
- Antigen synonyms:
- APP|CTFgamma|CVAP|protease nexin-II|ABETA|cerebral vascular amyloid peptide|beta-amyloid peptide|APPI|AD1|PN-II|Amyloid Precursor Protein|preA4|ABPP|alzheimer disease amyloid protein|Amyloid beta A4 protein|AAA|PN2|peptidase nexin-II
- APP|CTFgamma|CVAP|protease nexin-II|ABETA|cerebral vascular amyloid peptide|beta-amyloid peptide|APPI|AD1|PN-II|Amyloid Precursor Protein|preA4|ABPP|alzheimer disease amyloid protein|Amyloid beta A4 protein|AAA|PN2|peptidase nexin-II
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 4G8
- 4G8
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2b kappa
- IgG2b kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Format:
- Azide-free
- Azide-free
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- Phosphate-buffered solution (no preservatives or carrier proteins).
- Phosphate-buffered solution (no preservatives or carrier proteins).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- IHC-P, Direct ELISA, IHC-F, ICC, IP
- IHC-P, Direct ELISA, IHC-F, ICC, IP
Specifications
Frequently Bought Together