Order Entry
Switzerland
ContactUsLinkComponent
Anti-Histone H3 (phospho-Ser10) Mouse Monoclonal Antibody [clone: 11D8] (PE (Phycoerythrin))
Anti-Histone H3 (phospho-Ser10) Mouse Monoclonal Antibody [clone: 11D8] (PE (Phycoerythrin))
Supplier:  Biolegend
Certificates
Anti-Histone H3 (phospho-Ser10) Mouse Monoclonal Antibody [clone: 11D8] (PE (Phycoerythrin))
Supplier:  Biolegend

Specifications

  • Catalog No:
  • BLEG650807
  • BLEG650808
  • Antigen symbol:
  • Histone H3
  • Histone H3
  • Antigen synonyms:
  • Histone-H3|H3
  • Histone-H3|H3
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • 11D8
  • 11D8
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Modification:
  • phospho-Ser10
  • phospho-Ser10
  • Immunogen:
  • Modified synthetic peptide conjugated to KLH
  • Modified synthetic peptide conjugated to KLH
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
  • The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
  • Storage buffer:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Storage temperature:
  • 2…8 °C
  • 2…8 °C
  • Tested applications:
  • ICFC
  • ICFC

Specifications

Frequently Bought Together