Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (Brilliant Violet® 785)
Supplier: Chemicals
Certificates
About this item
Specifications
- Catalog No:
- BLEG300553
- XXXX300554
- Antigen symbol:
- CD4
- CD4
- Antigen synonyms:
- T4
- T4
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Brilliant Violet® 785
- Brilliant Violet® 785
- Clone:
- RPA-T4
- RPA-T4
- Reactivity:
- Chimpanzee, Human
- Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 785™ under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 785™ under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Specifications
Frequently Bought Together