Anti-CD3 Mouse Monoclonal Antibody [clone: UCHT1] (Brilliant Violet® 605)
- Catalog No:
- BLEG300460
- BLEG300459
- Antigen symbol:
- CD3
- CD3
- Antigen synonyms:
- T3|CD3ε
- T3|CD3ε
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Brilliant Violet® 605
- Brilliant Violet® 605
- Clone:
- UCHT1
- UCHT1
- Reactivity:
- Chimpanzee, Human
- Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 605™ under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 605™ under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Frequently Bought Together