Anti-CD196 (CCR6) Mouse Monoclonal Antibody [clone: G034E3] (PE (Phycoerythrin)/Dazzle™ 594)
- Catalog No:
- BLEG353429
- BLEG353430
- Antigen symbol:
- CD196 (CCR6)
- CD196 (CCR6)
- Antigen synonyms:
- STRL22|GPRCY4|GPR-CY4|CKR-L3|CKRL3|CMKBR6|CKR6|DRY-6|GPR29|BN-1|DCR2
- STRL22|GPRCY4|GPR-CY4|CKR-L3|CKRL3|CMKBR6|CKR6|DRY-6|GPR29|BN-1|DCR2
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)/Dazzle™ 594
- PE (Phycoerythrin)/Dazzle™ 594
- Clone:
- G034E3
- G034E3
- Reactivity:
- Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
- Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- CCR6-transfected cells
- CCR6-transfected cells
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE/Dazzle™ 594 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PE/Dazzle™ 594 under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Frequently Bought Together