Anti-CD324 (E-Cadherin) Rat Monoclonal Antibody [clone: DECMA-1] (PE (Phycoerythrin)/Cy7®)
- Catalog No:
- BLEG147310
- BLEG147309
- Antigen symbol:
- CD324 (E-Cadherin)
- CD324 (E-Cadherin)
- Antigen synonyms:
- E-Cadherin|Cadherin-1|CDH1|and UVO
- E-Cadherin|Cadherin-1|CDH1|and UVO
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)/Cy7®
- PE (Phycoerythrin)/Cy7®
- Clone:
- DECMA-1
- DECMA-1
- Reactivity:
- Pig, Cynomolgus monkey, Human, Dog, Mouse
- Pig, Cynomolgus monkey, Human, Dog, Mouse
- Host:
- Rat
- Rat
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- E-Cadherin extracellular domain
- E-Cadherin extracellular domain
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE/Cyanine7 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PE/Cyanine7 under optimal conditions.
- Concentration:
- 0.2 mg/ml
- 0.2 mg/ml
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Frequently Bought Together