Anti-FOXP3 Mouse Monoclonal Antibody [clone: 206D] (PE (Phycoerythrin)/Dazzle™ 594)
Catalog # BLEG320126
Supplier: Biolegend
Specifications
- Antigen symbol:FOXP3
- Clonality:Monoclonal
- Clone:206D
- Conjugation:PE (Phycoerythrin)/Dazzle™ 594
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Baboon,Cynomolgus monkey,Rhesus monkey,Pigtailed macaque,Human
- Antigen synonyms:Zinc finger protein JM2|JM2|Forkhead box protein P3|Scurfin|IPEX
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:2…8 °C
- Immunogen:Full-length FOXP3 protein
- Tested applications:ICFC
- Purification:The antibody was purified by affinity chromatography and conjugated with PE/Dazzle™ 594 under optimal conditions.
- Size:100 tests
- Pk:100 Tests