Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1]
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG363001
- BLEG363002
- Antigen symbol:
- CD19
- CD19
- Antigen synonyms:
- B-lymphocyte Antigen CD19|T-cell Surface Antigen Leu-12|CVID3|CD19 Antigen|CD19 Molecule|B-Lymphocytes Surface Antigen B4|Differentiation Antigen CD19|B4
- B-lymphocyte Antigen CD19|T-cell Surface Antigen Leu-12|CVID3|CD19 Antigen|CD19 Molecule|B-Lymphocytes Surface Antigen B4|Differentiation Antigen CD19|B4
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- SJ25C1
- SJ25C1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- NALM1 + NALM16 cells
- NALM1 + NALM16 cells
- Format:
- Purified
- Purified
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC, IHC-F
- FC, IHC-F
Specifications
Frequently Bought Together