Anti-CD45RA Mouse Monoclonal Antibody [clone: HI100] (Brilliant Violet® 570)
Catalog # BLEG304132
Supplier: Biolegend
Specifications
- Antigen-Symbol:CD45RA
- Klonalität:Monoklonal
- Klon:HI100
- Konjugation:Brilliant Violet® 570
- Durchflusszytometrie:Yes
- Wirt:Mouse
- Isotyp:IgG2b kappa
- Reaktivität:Chimpanzee,Human
- Antigen-Synonyme:PTPRC|GP180|T200|L-CA|LY5|LCA
- Lagerungspuffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Lagertemperatur:2…8 °C
- Validierte Anwendungen:FC
- Reinigung:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 570™ under optimal conditions.
- Größe:100 tests
- VE:100 Tests