Anti-CD25 Mouse Monoclonal Antibody [clone: BC96] (Brilliant Violet® 650)
Catalog # BLEG302634
Supplier: Biolegend
Specifications
- Antigen symbol:CD25
- Clonality:Monoclonal
- Clone:BC96
- Conjugation:Brilliant Violet® 650
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Baboon,Cynomolgus monkey,Rhesus monkey,Pigtailed macaque,Chimpanzee,Human
- Antigen synonyms:IL-2R α chain|Tac|p55|Low affinity IL-2R
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:2…8 °C
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 650™ under optimal conditions.
- Size:100 tests
- Pk:100 Tests