Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD20 Mouse Monoclonal Antibody [clone: 2H7] (Brilliant Violet® 650)
Anti-CD20 Mouse Monoclonal Antibody [clone: 2H7] (Brilliant Violet® 650)
Catalog # BLEG302336
Supplier:  Biolegend
Anti-CD20 Mouse Monoclonal Antibody [clone: 2H7] (Brilliant Violet® 650)
Catalog # BLEG302336
Supplier:  Biolegend

Specifications

  • Antigen symbol:
    CD20
  • Clonality:
    Monoclonal
  • Clone:
    2H7
  • Conjugation:
    Brilliant Violet® 650
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Baboon,
    Capuchin monkey,
    Cynomolgus monkey,
    Rhesus monkey,
    Pigtailed macaque,
    Chimpanzee,
    Human,
    Squirrel monkey
  • Antigen synonyms:
    Bp35|B1
  • Storage buffer:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Storage temperature:
    2…8 °C
  • Immunogen:
    Human tonsillar B cells
  • Tested applications:
    FC
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 650™ under optimal conditions.
  • Size:
    100 tests
  • Pk:
    100 Tests

Specifications

About this item