Anti-CD14 Mouse Monoclonal Antibody [clone: M5E2] (Brilliant Violet® 650)
Catalog # BLEG301836
Supplier: Biolegend
Specifications
- Antigen symbol:CD14
- Clonality:Monoclonal
- Clone:M5E2
- Conjugation:Brilliant Violet® 650
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Capuchin monkey,Squirrel monkey,African green monkey,Cynomolgus monkey,Common marmoset,Rhesus monkey,Cotton-topped tamarin,Pigtailed macaque,Chimpanzee,Human,Bovine,Dog
- Antigen synonyms:LPS receptor
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:2…8 °C
- Immunogen:Full-length human CD14 protein
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 650™ under optimal conditions.
- Size:100 tests
- Pk:100 Tests