Anti-CD144 (VE-Cadherin) Mouse Monoclonal Antibody [clone: BV9] (Alexa Fluor® 647)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG348514
- BLEG348513
- Antigen-Symbol:
- CD144 (VE-Cadherin)
- CD144 (VE-Cadherin)
- Antigen-Synonyme:
- CDH5|VE-Cadherin|cadherin-5
- CDH5|VE-Cadherin|cadherin-5
- Klonalität:
- Monoklonal
- Monoklonal
- Konjugation:
- Alexa Fluor® 647
- Alexa Fluor® 647
- Klon:
- BV9
- BV9
- Reaktivität:
- Human
- Human
- Wirt:
- Mouse
- Mouse
- Isotyp:
- IgG2a kappa
- IgG2a kappa
- ImmunoChemie:
- Yes
- Yes
- Durchflusszytometrie:
- Yes
- Yes
- Reinigung:
- The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 647 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 647 under optimal conditions.
- Lagerungspuffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Lagertemperatur:
- 2…8 °C
- 2…8 °C
- Validierte Anwendungen:
- FC, IHC-P
- FC, IHC-P
Specifications
Frequently Bought Together