Order Entry
Switzerland
ContactUsLinkComponent
Anti-IFN-gamma Mouse Monoclonal Antibody [clone: 4S.B3] (Brilliant Violet® 650)
Anti-IFN-gamma Mouse Monoclonal Antibody [clone: 4S.B3] (Brilliant Violet® 650)
Catalog # BLEG502538
Supplier:  Biolegend
Anti-IFN-gamma Mouse Monoclonal Antibody [clone: 4S.B3] (Brilliant Violet® 650)
Catalog # BLEG502538
Supplier:  Biolegend

Specifications

  • Antigen symbol:
    IFN-gamma
  • Clonality:
    Monoclonal
  • Clone:
    4S.B3
  • Conjugation:
    Brilliant Violet® 650
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Baboon,
    Cynomolgus monkey,
    Rhesus monkey,
    Chimpanzee,
    Human
  • Antigen synonyms:
    Type II interferon|T cell interferon|Macrophage-activating factor (MAF)|IFN-gamma|IFN-g|Interferon-γ|Immune interferon
  • Storage buffer:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Storage temperature:
    2…8 °C
  • Immunogen:
    Partially purified, native human IFN-γ
  • Tested applications:
    ICFC
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 650™ under optimal conditions.
  • Size:
    100 tests
  • Pk:
    100 Tests

Specifications

About this item