Anti-CD39 Mouse Monoclonal Antibody [clone: A1] (Brilliant Violet® 510)
BLEG328219
- Antigen symbol:CD39
- Clonality:Monoclonal
- Clone:A1
- Conjugation:Brilliant Violet® 510
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Rhesus monkey,Human
- Antigen synonyms:E-ATPDase|NTPDase-1|ecto-apyrase|Ec3.6.1.5|gp80
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:2…8 °C
- Immunogen:PHA activated human lymphocytes
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 510™ under optimal conditions.
- Size:25 tests
- Pk:25 Tests