Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (Alexa Fluor® 594)
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (Alexa Fluor® 594)
Catalog # BLEG300544
Supplier:  Biolegend
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (Alexa Fluor® 594)
Catalog # BLEG300544
Supplier:  Biolegend

Specifications

  • Antigen symbol:
    CD4
  • Clonality:
    Monoclonal
  • Clone:
    RPA-T4
  • Conjugation:
    Alexa Fluor® 594
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Chimpanzee,
    Human
  • Antigen synonyms:
    T4
  • Storage buffer:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Storage temperature:
    2…8 °C
  • Concentration:
    0.5 mg/ml
  • Tested applications:
    IF, FC
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 594 under optimal conditions.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item