Anti-CD120b Rat Monoclonal Antibody [clone: 3G7A02] (Ultra-LEAF™)
BLEG358408
- Antigen symbol:CD120b
- Clonality:Monoclonal
- Clone:3G7A02
- Conjugation:Ultra-LEAF™
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Human
- Neutralisation:Yes
- Antigen synonyms:TBPII|TNFRSF1B|p75|p75TNFR|TNFR2|TNFR80
- Storage buffer:0.2 µm filtered in phosphate-buffered solution, pH 7.2, containing no preservative. Endotoxin level is <0.01 EU/µg of the protein (<0.001 ng/µg of the protein) as determined by the LAL test.
- Storage temperature:2…8 °C
- Immunogen:Partial CD120b recombinant protein expressed in 293E cells (22-211aa).
- Tested applications:Neut, FC
- Purification:The Ultra-LEAF™ (Low Endotoxin, Azide-Free) antibody was purified by affinity chromatography.
- Size:500 µg
- Pk:500 µG