Anti-CD19 Rat Monoclonal Antibody [clone: 6D5] (Brilliant Violet® 605)
Catalog # BLEG115540
Supplier: Biolegend
Specifications
- Antigen symbol:CD19
- Clonality:Monoclonal
- Clone:6D5
- Conjugation:Brilliant Violet® 605
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen synonyms:B4
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:2…8 °C
- Concentration:0.2 mg/ml
- Immunogen:Mouse CD19-expressing K562 human erythroleukemia cells
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 605™ under optimal conditions.
- Size:50 µg
- Pk:50 µG