Anti-CD4 Mouse Monoclonal Antibody [clone: OKT4] (Brilliant Violet® 711)
Katalognr. BLEG317440
Anbieter: Biolegend
Technische Daten
- Antigen symbol:CD4
- Clonality:Monoclonal
- Clone:OKT4
- Conjugation:Brilliant Violet® 711
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Cynomolgus monkey,Rhesus monkey,Chimpanzee,Human
- Antigen synonyms:T4
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:2…8 °C
- Immunogen:Human peripheral T cells
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711™ under optimal conditions.
- Size:100 tests
- Pk:100 Tests