Anti-GATA3 Mouse Monoclonal Antibody [clone: 16E10A23] (PE (Phycoerythrin))
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG653804
- BLEG653803
- Antigen symbol:
- GATA3
- GATA3
- Antigen synonyms:
- trans-acting T-cell-specific transcription factor GATA-3|GATA-binding factor 3
- trans-acting T-cell-specific transcription factor GATA-3|GATA-binding factor 3
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- 16E10A23
- 16E10A23
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Partial human GATA3 recombinant protein (1-258 aa).
- Partial human GATA3 recombinant protein (1-258 aa).
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- ICFC
- ICFC
Specifications
Frequently Bought Together