Anti-CD163 Mouse Monoclonal Antibody [clone: GHI/61] (Brilliant Violet® 421)
Fournisseur: Biolegend
Certificats
À propos de cet article
Spécifications
- Catalog No:
- BLEG333611
- BLEG333612
- Antigen symbol:
- CD163
- CD163
- Antigen synonyms:
- Hemoglobin/Haptoglobin Complex Receptor|M130|ED2(rat)|GHI/61|RM3/1|p155|macrophage-associated antigen
- Hemoglobin/Haptoglobin Complex Receptor|M130|ED2(rat)|GHI/61|RM3/1|p155|macrophage-associated antigen
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Brilliant Violet® 421
- Brilliant Violet® 421
- Clone:
- GHI/61
- GHI/61
- Reactivity:
- African green monkey, Cynomolgus monkey, Rhesus monkey, Human
- African green monkey, Cynomolgus monkey, Rhesus monkey, Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Spécifications
Frequently Bought Together