Anti-CD360 (IL-21R) Mouse Monoclonal Antibody [clone: 17A12] (PE (Phycoerythrin))
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG359506
- BLEG359505
- Antigen symbol:
- CD360 (IL-21R)
- CD360 (IL-21R)
- Antigen synonyms:
- IL-21 Receptor|MGC10967|novel interleukin receptor (NILR)
- IL-21 Receptor|MGC10967|novel interleukin receptor (NILR)
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- 17A12
- 17A12
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Human IL-21R expressing mouse M1-T22 cell line.
- Human IL-21R expressing mouse M1-T22 cell line.
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Specifications
Frequently Bought Together