Anti-CD304 (Neuropilin-1) Mouse Monoclonal Antibody [clone: 12C2] (FITC (Fluorescein Isothiocyanate))
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG354511
- BLEG354512
- Antigen symbol:
- CD304 (Neuropilin-1)
- CD304 (Neuropilin-1)
- Antigen synonyms:
- BDCA-4
- BDCA-4
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- 12C2
- 12C2
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- CD304-Fc Fusion protein
- CD304-Fc Fusion protein
- Purification:
- The antibody was purified by affinity chromatography and conjugated with FITC under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with FITC under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Specifications
Frequently Bought Together