Anti-CD185 (CXCR5) Mouse Monoclonal Antibody [clone: J252D4] (Brilliant Violet® 421)
- Catalog No:
- BLEG356920
- BLEG356919
- Antigen symbol:
- CD185 (CXCR5)
- CD185 (CXCR5)
- Antigen synonyms:
- BLR1|MDR15
- BLR1|MDR15
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Brilliant Violet® 421
- Brilliant Violet® 421
- Clone:
- J252D4
- J252D4
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Human CXCR5-transfected cells
- Human CXCR5-transfected cells
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Frequently Bought Together