Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD15 (SSEA-1) Mouse Monoclonal Antibody [clone: W6D3] (Brilliant Violet® 605)
Anti-CD15 (SSEA-1) Mouse Monoclonal Antibody [clone: W6D3] (Brilliant Violet® 605)
Catalog # BLEG323032
Supplier:  Biolegend
Anti-CD15 (SSEA-1) Mouse Monoclonal Antibody [clone: W6D3] (Brilliant Violet® 605)
Catalog # BLEG323032
Supplier:  Biolegend

Specifications

  • Antigen symbol:
    CD15 (SSEA-1)
  • Clonality:
    Monoclonal
  • Clone:
    W6D3
  • Conjugation:
    Brilliant Violet® 605
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Antigen synonyms:
    LNFP III|X-hapten|FUC-TIV|ELFT|3-FL|SSEA1|Lewis X|SSEA-1|FUT4|FCT3A|FUTIV|Lex |3-FAL
  • Storage buffer:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Storage temperature:
    2…8 °C
  • Immunogen:
    WERI-RB-1 retinoblastoma cell line
  • Tested applications:
    FC
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 605™ under optimal conditions.
  • Size:
    100 tests
  • Pk:
    100

Specifications

About this item