Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (Brilliant Violet® 605)
Catalog # BLEG302244
Anbieter: Biolegend
Technische Daten
- Antigen synonyms:B4
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:2…8 °C
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 605™ under optimal conditions.
- Size:100 tests
- Pk:100 Tests